missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Antibody (5H10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11310AF488
Additional Details : Weight : 0.00970kg
Description
Desmoglein-3 Monoclonal antibody specifically detects Desmoglein-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
Desmoglein-3 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
5H10 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA | |
Recombinant human DSG3 extracellular protein fragment | |
0.1 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction | |
1830 | |
Store at 4°C in the dark. | |
IgG1 κ |