Få mere at vide
Desmethylthiamine Chloride Hydrochloride, Mikromol™
Artikelnummer. 30049422
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30049422 25 mg
1 options
Artikelnummer. 30049422 Leverandør Mikromol Leverandørnr. MM0049.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Desmethylthiamine Chloride Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2'-Nor Thiamine Hydrochloride Salt
CAS 49614-72-4
Analyt- eller komponentnavne 2'-Nor Thiamine Hydrochloride Salt
Forsendelsestilstand Room Temperature
Molekylær formel C11 H15 N4 O S . Cl . Cl H
Certificeringer/compliance ISO 9001
Synonym Thiazolium, 3-[(4-amino-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, hydrochloride (1:1:1), Thiazolium, 3-[(4-amino-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-, chloride, monohydrochloride (9CI)
InChI formel InChI=1S/C11H15N4OS.2ClH/c1-8-10(2-3-16)17-7-15(8)5-9-4-13-6-14-11(9)12;;/h4,6-7,16H,2-3,5H2,1H3,(H2,12,13,14);2*1H/q+1;;/p-1
SMIL Cl.[Cl-].Cc1c(CCO)sc[n+]1Cc2cncnc2N
IUPAC navn 2-[3-[(4-aminopyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
Molekylvægt (g/mol) 323.24
Mængde 25 mg
Formel vægt 322.0422 g/mol