Få mere at vide
Desmethyl Thiosildenafil-d8, TRC
Artikelnummer. 30366287
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30366287 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366287 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29435210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desmethyl Thiosildenafil-d8
CAS 1215321-44-0
Molekylær formel C21H20D8N6O3S2
Mængde 10 mg
Synonym 5-[2-Ethoxy-5-(1-piperazinyl-d8-sulfonyl)phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione
InChI formel InChI=1S/C21H28N6O3S2/c1-4-6-16-18-19(26(3)25-16)21(31)24-20(23-18)15-13-14(7-8-17(15)30-5-2)32(28,29)27-11-9-22-10-12-27/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,23,24,31)/i9D2,10D2,11D2,12D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C1([2H])[2H])S(=O)(=O)c2ccc(OCC)c(c2)C3=NC(=S)c4c(N3)c(CCC)nn4C
IUPAC navn 5-[2-ethoxy-5-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidine-7-thione
Molekylvægt (g/mol) 484.66
Formel vægt 484.2166