Få mere at vide
Desmethyl-O-Benzyl Apixaban, TRC
Artikelnummer. 30359102
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359102 100 mg
30364719 50 mg
30365830 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30359102 Leverandør Toronto Research Chemicals Leverandørnr. TRCD119845100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1620494-31-6
Molekylær formel C31 H29 N5 O4
Molekylvægt (g/mol) 535.593
Synonym 1-(4-(Benzyloxy)phenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxamide
IUPAC navn 7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1-(4-phenylmethoxyphenyl)-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxamide
SMIL NC(=O)c1nn(c2ccc(OCc3ccccc3)cc2)c4C(=O)N(CCc14)c5ccc(cc5)N6CCCCC6=O
Tekniske data
Kemisk navn eller materiale Desmethyl-O-Benzyl Apixaban
Mængde 100 mg
InChI formel InChI=1S/C31H29N5O4/c32-30(38)28-26-17-19-35(23-11-9-22(10-12-23)34-18-5-4-8-27(34)37)31(39)29(26)36(33-28)24-13-15-25(16-14-24)40-20-21-6-2-1-3-7-21/h1-3,6-7,9-16H,4-5,8,17-20H2,(H2,32,38)
Formel vægt 535.222