Få mere at vide
Desmethyl Icaritin, TRC
Artikelnummer. 30357515
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357515 10 mg
30350272 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30357515 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29176010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 28610-31-3
Molekylær formel C20 H18 O6
Molekylvægt (g/mol) 354.35
Synonym 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, 3,4',5,7-Tetrahydroxy-8-(3-methyl-2-butenyl)flavone, 4'-Desmethylicaritin, 8-Prenylkaempferol, De-O-methylanhydroicaritin, Noranhydroicaritin
IUPAC navn 3,5,7-trihydroxy-2-(4-hydroxyphenyl)-8-(3-methylbut-2-enyl)chromen-4-one
SMIL CC(=CCc1c(O)cc(O)c2C(=O)C(=C(Oc12)c3ccc(O)cc3)O)C
Tekniske data
Kemisk navn eller materiale desmethyl Icaritin
Mængde 10 mg
InChI formel InChI=1S/C20H18O6/c1-10(2)3-8-13-14(22)9-15(23)16-17(24)18(25)19(26-20(13)16)11-4-6-12(21)7-5-11/h3-7,9,21-23,25H,8H2,1-2H3
Formel vægt 354.1103