Få mere at vide
Desmethyl Cyclobenzaprine Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desmethylcyclobenzaprine HCl |
| CAS | 438-59-5 |
| Molekylær formel | C19 H19 N . Cl H |
| Mængde | 10 mg |
| Synonym | 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (1:1), 1-Propanamine, 3-(5H-dibenzo[a,d]cyclohepten-5-ylidene)-N-methyl-, hydrochloride (9CI), 5H-Dibenzo[a,d]cycloheptene-δ5,γ-propylamine, N-methyl-, hydrochloride (7CI,8CI), 10,11-Didehydronortriptyline hydrochloride, N 7068 Hydrochloride, Norproheptatriene hydrochloride, Ro 4-6011, 3-(5H-Dibenzo-[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine hydrochloride, N-Desmethylcyclobenzaprine hydrochloride, Norcyclobenzaprine hydrochloride |
| InChI formel | InChI=1S/C19H19N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-13,20H,6,14H2,1H3;1H |
| SMIL | Cl.CNCCC=C1c2ccccc2C=Cc3ccccc13 |
| IUPAC navn | 3-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Molekylvægt (g/mol) | 297.82 |
| Formel vægt | 297.1284 |