Få mere at vide
Desmethyl Atomoxetine-d7 Hydrochloride Salt, TRC
Artikelnummer. 30362250
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30362250 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362250 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2908271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desmethyl Atomoxetine-d7 Hydrochloride Salt
CAS 1189961-95-2
Molekylær formel C16H13D7ClNO
Mængde 1 mg
Synonym gamma-(2-Methylphenoxy-d7)benzenepropanamine Hydrochloride Salt
InChI formel InChI=1S/C16H19NO.ClH/c1-13-7-5-6-10-15(13)18-16(11-12-17)14-8-3-2-4-9-14;/h2-10,16H,11-12,17H2,1H3;1H/i1D3,5D,6D,7D,10D;
SMIL Cl.[2H]c1c([2H])c([2H])c(c(OC(CCN)c2ccccc2)c1[2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 284.83