Få mere at vide
Desmethyl Acridinium C2 NHS Ester, TRC
Artikelnummer. 30359479
Skift visning
Mængde:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359479 1 mg
30353669 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30359479 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2907001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desmethyl Acridinium NHS Ester
CAS 87198-87-6
Molekylær formel C27 H20 N2 O6
Mængde 1 mg
InChI formel InChI=1S/C27H20N2O6/c30-23-14-15-24(31)29(23)35-25(32)16-11-17-9-12-18(13-10-17)34-27(33)26-19-5-1-3-7-21(19)28-22-8-4-2-6-20(22)26/h1-10,12-13H,11,14-16H2
SMIL O=C(CCc1ccc(OC(=O)c2c3ccccc3nc4ccccc24)cc1)ON5C(=O)CCC5=O
IUPAC navn [4-[3-(2,5-dioxopyrrolidin-1-yl)oxy-3-oxopropyl]phenyl] acridine-9-carboxylate
Molekylvægt (g/mol) 468.46
Formel vægt 468.1321