Få mere at vide
Desloratadine-d4, TRC
Artikelnummer. 30353152
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30353152 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353152 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29025225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desloratadine-d4
CAS 381727-29-3
Molekylær formel C19 2H4 H15 Cl N2
Mængde 25 mg
Synonym Piperidine-2,2,6,6-d4, 4-(8-chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)- (9CI), 8-Chloro-6,11-dihydro-11-(4-piperidinylidene-2,2,6,6-d4)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, 8-Chloro-11-(4-piperidylidene-2,2,6,6-d4)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Descarboethoxyloratadine-d4, Desloratadine-d4, Desloratadine-D4 (piperidine-2,2,6,6-D4)
InChI formel InChI=1S/C19H19ClN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2/i10D2,11D2
SMIL [2H]C1([2H])CC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC([2H])([2H])N1
IUPAC navn 8-chloro-11-(2,2,6,6-tetradeuteriopiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
Molekylvægt (g/mol) 314.85
Formel vægt 314.1488