Få mere at vide
Desloratadine-d1 N-Oxide, TRC
Artikelnummer. 30361478
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30361478 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361478 Leverandør Toronto Research Chemicals Leverandørnr. TRCD290262100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desloratadine-d1 N-Oxide
CAS 1794817-18-7
Molekylær formel C19H18DClN2O
Mængde 100 mg
Synonym 8-Chloro-6,11-dihydro-11-(4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine-d 1-Oxide
InChI formel InChI=1S/C19H19ClN2O/c20-16-5-6-17-15(12-16)4-3-14-2-1-11-22(23)19(14)18(17)13-7-9-21-10-8-13/h1-2,5-6,11-12,21H,3-4,7-10H2/i11D
SMIL [2H]c1ccc2CCc3cc(Cl)ccc3C(=C4CCNCC4)c2[n+]1[O-]
IUPAC navn 8-chloro-2-deuterio-1-oxido-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[3,4-b]pyridin-1-ium
Molekylvægt (g/mol) 327.83
Formel vægt 327.1249