Få mere at vide
Desisopropyl Atrazine-d5, TRC
Artikelnummer. 30349386
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30349386 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349386 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2901522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Atrazine-desisopropyl D5 (ethylamino D5)
CAS 1189961-78-1
Molekylær formel C5 H8 Cl N5
Mængde 2.5 mg
Synonym 6-Chloro-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
InChI formel InChI=1S/C5H8ClN5/c1-2-8-5-10-3(6)9-4(7)11-5/h2H2,1H3,(H3,7,8,9,10,11)/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Nc1nc(N)nc(Cl)n1
IUPAC navn 6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 178.63
Formel vægt 178.0782