Få mere at vide
Deshydroxyethylopipramol Dihydrochloride (5-(3-Piperazin-1-yl-propyl)-5H-dibenzo[b,f]azepine Dihydrochloride), LoGiCal
Artikelnummer. 30458524
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458524 10 mg
1 options
Artikelnummer. 30458524 Leverandør LoGiCal Leverandørnr. LGCFOR0422.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deshydroxyethylopipramol Dihydrochloride
CAS 2727065-82-7
Analyt- eller komponentnavne Deshydroxyethylopipramol Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C21 H25 N3 . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym 5-(3-Piperazin-1-yl-propyl)-5H-dibenzo[b,f]azepine Dihydrochloride, Deshydroxyethylopipramol Dihydrochloride
InChI formel InChI=1S/C21H25N3.2ClH/c1-3-8-20-18(6-1)10-11-19-7-2-4-9-21(19)24(20)15-5-14-23-16-12-22-13-17-23;;/h1-4,6-11,22H,5,12-17H2;2*1H
Løsningstype Neat
SMIL Cl.Cl.C(CN1CCNCC1)CN2c3ccccc3C=Cc4ccccc24
IUPAC navn 11-(3-piperazin-1-ylpropyl)benzo[b][1]benzazepine;dihydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 392.37
Mængde 10 mg
Formel vægt 391.1582 g/mol