Få mere at vide
Desfuroyl Ceftiofur Dimer (>90%), TRC
Artikelnummer. 30132915
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132915 5 mg
30135919 10 mg
30012208 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132915 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2904335MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120905-00-2
Molekylær formel C28 H28 N10 O10 S6
Molekylvægt (g/mol) 856.97
Synonym (6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-3-[[[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-(methoxyimino)acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]dithio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
IUPAC navn (6R,7R)-7-[[(2Z)-2-(2-aminothiazol-4-yl)-2-methoxyimino-acetyl]amino]-3-[[[(6R,7R)-7-[[(2Z)-2-(2-aminothiazol-4-yl)-2-methoxyimino-acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyldisulfanyl]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMIL CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSSCC3=C(N4[C@H](SC3)[C@H](NC(=O)\C(=N/OC)\c5csc(N)n5)C4=O)C(=O)O)\c6csc(N)n6
Tekniske data
Kemisk navn eller materiale Desfuroyl Ceftiofur Dimer
Mængde 5 mg
InChI formel InChI=1S/C28H28N10O10S6/c1-47-35-13(11-7-51-27(29)31-11)19(39)33-15-21(41)37-17(25(43)44)9(3-49-23(15)37)5-53-54-6-10-4-50-24-16(22(42)38(24)18(10)26(45)46)34-20(40)14(36-48-2)12-8-52-28(30)32-12/h7-8,15-16,23-24H,3-6H2,1-2H3,(H2,29,31)(H2,30,32)(H,33,39)(H,34,40)(H,43,44)(H,45,46)/b35-13-,36-14-/t15-,16-,23-,24-/m1/s1
Formel vægt 856.0314