Få mere at vide
Tocris Bioscience™ Desformylflustrabromine hydrochloride
GREENER_CHOICE
Artikelnummer. 18045062
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18045062 50 mg
18035682 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18045062 Leverandør Tocris Bioscience™ Leverandørnr. 3328/50
Positive allosteric modulator of α4β2; also muscle-type nAChR inhibitor
Positive allosteric modulator of nicotinic α4β2 receptors; selectively increases the ionic current through α4β2 in the presence of ACh. Displays 14.7-fold selectivity for α4β2 over homomeric (α7) receptors. Moderately cytotoxic in HCT-116 cells. Also inhibits human muscle (αβεδ) and Torpedo (αβγδ) nAChRs (IC50 values are 1.0 and 0.1 μM, respectively) by binding in the ion channel.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 951322-11-5
Molekylær formel C16H22BrClN2
Tekniske data
Kemisk navn eller materiale Desformylflustrabromine hydrochloride
Mængde 50 mg
Formel vægt 357.72
Renhed >98%
Molekylvægt (g/mol) 357.72