Få mere at vide
Desethyloxybutynin Hydrochloride, Mikromol™
Artikelnummer. 17942296
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17942296 25 mg
17982036 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17942296 Leverandør Mikromol Leverandørnr. MM0047.110025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Desethyloxybutynin Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desethyloxybutynin Hydrochloride
CAS 81039-77-2
Analyt- eller komponentnavne Desethyloxybutynin Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C20 H27 N O3 . Cl H
Certificeringer/compliance ISO 17025
Synonym Desethyloxybutynin Hydrochloride
InChI formel InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H
SMIL Cl.CCNCC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
IUPAC navn 4-(ethylamino)but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride
Molekylvægt (g/mol) 365.89
Mængde 25 mg
Formel vægt 365.1758 g/mol