Få mere at vide
Desethylene Norfloxacin Hydrochloride, TRC
Artikelnummer. 30011712
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011712 25 mg
30006264 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011712 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28976025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797116-86-9
Molekylær formel C14 H16 F N3 O3 . Cl H
Molekylvægt (g/mol) 329.75
Synonym 3-Quinolinecarboxylic acid, 7-[(2-aminoethyl)amino]-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-, hydrochloride (1:1), 7-[(2-Aminoethyl)amino]-1-ethyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride, 7-((2-Aminoethyl)amino)-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Desethylene Norfloxacin hydrochloride
IUPAC navn 7-(2-aminoethylamino)-1-ethyl-6-fluoro-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMIL Cl.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(NCCN)cc12
Tekniske data
Kemisk navn eller materiale 7-[(2-Aminoethyl)amino]-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid Hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C14H16FN3O3.ClH/c1-2-18-7-9(14(20)21)13(19)8-5-10(15)11(6-12(8)18)17-4-3-16;/h5-7,17H,2-4,16H2,1H3,(H,20,21);1H
Formel vægt 329.0942