Få mere at vide
Desethylclomifene Hydrochloride, LoGiCal
Artikelnummer. 30461125
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461125 10 mg
1 options
Artikelnummer. 30461125 Leverandør LoGiCal Leverandørnr. LGCFOR0650.03
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desethylclomifene Hydrochloride
CAS 1310815-19-0
Analyt- eller komponentnavne Desethylclomifene Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C24 H24 Cl N O . Cl H
Certificeringer/compliance ISO 9001
Synonym Desethylclomifene Hydrochloride
InChI formel InChI=1S/C24H24ClNO.ClH/c1-2-26-17-18-27-22-15-13-20(14-16-22)23(19-9-5-3-6-10-19)24(25)21-11-7-4-8-12-21;/h3-16,26H,2,17-18H2,1H3;1H
Løsningstype Neat
SMIL Cl.CCNCCOc1ccc(cc1)C(=C(Cl)c2ccccc2)c3ccccc3
IUPAC navn 2-[4-(2-chloro-1,2-diphenylethenyl)phenoxy]-N-ethylethanamine;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 414.37
Mængde 10 mg
Formel vægt 413.1313 g/mol