Få mere at vide
Desethylatrazine-d7 (iso-propyl-d7), CDN
Artikelnummer. 30319421
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319421 0.01 g
30317639 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319421 Leverandør CDN Leverandørnr. CDND56390.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Amino-4-(isopropyl-d7-amino)-6-chloro-triazine
CAS 1216649-31-8
Molekylær formel C6 D7 H3 Cl N5
Mængde 0.01 g
Synonym Atrazine-desethyl D7 (Isopropyl D7), 6-Chloro-N2-[1-(methyl-d3)ethyl-1,2,2,2-d4]-1,3,5-triazine-2,4-diamine, 6-Chloro-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine, D7 deethylatrazine
InChI formel InChI=1 S/C6H10ClN5/c1-3(2)9-6-11-4(7)10-5(8)12-6/h3H,1-2H3,(H3,8,9,10,11,12)/i1D3,2D3,3 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(N)nc(Cl)n1)C([2 H])([2 H])[2 H]
IUPAC navn 6-chloro-2-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 194.6732
Formel vægt 194.1064 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity