Få mere at vide
Desethylamiodarone Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desethylamiodarone HCl |
| CAS | 96027-74-6 |
| Analyt- eller komponentnavne | N-Desethylamiodarone HCl |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H25 I2 N O3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | (2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone Hydrochloride (Desethylamiodarone Hydrochloride), Amiodarone Hydrochloride Imp. B (EP), Amiodarone Imp. B (EP), (2-Butylbenzofuran-3-yl)[4-[2-(ethylamino)ethoxy]-3,5-diiodophenyl]methanone Hydrochloride, Amiodarone Hydrochloride Imp. B (EP) as Hydrochloride, Desethylamiodarone Hydrochloride, Amiodarone Hydrochloride Impurity B as Hydrochloride, Amiodarone Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C23H25I2NO3.ClH/c1-3-5-9-20-21(16-8-6-7-10-19(16)29-20)22(27)15-13-17(24)23(18(25)14-15)28-12-11-26-4-2;/h6-8,10,13-14,26H,3-5,9,11-12H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCNCC)c(I)c3 |
| Vis mere |