Få mere at vide
Desethyl Sildenafil-d3, TRC
Artikelnummer. 30358548
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30358548 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358548 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28960210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desethyl Sildenafil-d3
CAS 1346603-75-5
Molekylær formel C20H23D3N6O4S
Mængde 10 mg
Synonym 5-[2-Hydroxy-5-(4-methyl-d3-piperazinylsulphonyl)phenyl]1-methyl-3-n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyramidin-7-one, 5-[2-Hydroxy-5-[[4-(methyl-d3)-1-piperazinyl]sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one
InChI formel InChI=1S/C20H26N6O4S/c1-4-5-15-17-18(25(3)23-15)20(28)22-19(21-17)14-12-13(6-7-16(14)27)31(29,30)26-10-8-24(2)9-11-26/h6-7,12,21H,4-5,8-11H2,1-3H3,(H,22,28)/b19-14+/i2D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)S(=O)(=O)C2=C\C(=C\3/NC(=O)c4c(N3)c(CCC)nn4C)\C(=O)C=C2
IUPAC navn (5E)-1-methyl-5-[6-oxo-3-[4-(trideuteriomethyl)piperazin-1-yl]sulfonylcyclohexa-2,4-dien-1-ylidene]-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
Molekylvægt (g/mol) 449.52
Formel vægt 449.1925