Få mere at vide
Deserpidine, LoGiCal
Artikelnummer. 30459900
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30459900 5 mg
1 options
Artikelnummer. 30459900 Leverandør LoGiCal Leverandørnr. LGCFOR1654.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deserpidine
CAS 131-01-1
Analyt- eller komponentnavne Deserpidine
Forsendelsestilstand Room Temperature
Molekylær formel C32 H38 N2 O8
Certificeringer/compliance ISO 9001
Synonym 3β,20α-Yohimban-16β-carboxylic acid, 18β-hydroxy-17α-methoxy-, methyl ester, 3,4,5-trimethoxybenzoate (ester) (8CI), Deserpidic acid, methyl ester, 3,4,5-trimethoxybenzoate (6CI), Deserpidine (7CI), Benz[g]indolo[2,3-a]quinolizine, yohimban-16-carboxylic acid deriv., 11-Demethoxyreserpine, 11-Desmethoxyreserpine, Canescin, Canescine, Canescine (Rauwolfia), Deserpic acid, methyl ester, 3,4,5-trimethoxybenzoate, Deserpidin, Harmonyl, NSC 72138, Raunormin, Raunormine, Recanescin, Recanescine, Reserpidine, Yohimban-16-carboxylic acid, 17-methoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]-, methyl ester, (3β,16β,17α,18β,20α)-
InChI formel InChI=1S/C32H38N2O8/c1-37-24-12-17(13-25(38-2)29(24)39-3)31(35)42-26-14-18-16-34-11-10-20-19-8-6-7-9-22(19)33-28(20)23(34)15-21(18)27(30(26)40-4)32(36)41-5/h6-9,12-13,18,21,23,26-27,30,33H,10-11,14-16H2,1-5H3/t18-,21+,23-,26-,27+,30+/m1/s1
Løsningstype Neat
SMIL CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5ccccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6
IUPAC navn methyl (1R,15S,17R,18R,19S,20S)-18-methoxy-17-(3,4,5-trimethoxybenzoyl)oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
Industry Type Clinical Testing
Molekylvægt (g/mol) 578.65
Mængde 5 mg
Formel vægt 578.2628 g/mol