Få mere at vide
Desdiacetyl-8-oxo Famciclovir-d4, TRC
Artikelnummer. 30355053
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30355053 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355053 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28893210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desdiacetyl-8-oxo Famciclovir-d4
CAS 1346603-55-1
Molekylær formel C10H11D4N5O3
Mængde 10 mg
Synonym 2-Amino-7,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-8H-purin-8-one-d4, BRL 48959-d4, 8-Oxo-6-deoxypenciclovir-d4
InChI formel InChI=1S/C10H15N5O3/c11-9-12-3-7-8(14-9)15(10(18)13-7)2-1-6(4-16)5-17/h3,6,16-17H,1-2,4-5H2,(H,13,18)(H2,11,12,14)/i1D2,2D2
SMIL [2H]C([2H])(C(CO)CO)C([2H])([2H])N1C(=NC2=CNC(=N)N=C12)O
IUPAC navn 2-[1,1,2,2-tetradeuterio-2-(8-hydroxy-2-imino-1H-purin-9-yl)ethyl]propane-1,3-diol
Molekylvægt (g/mol) 257.28
Formel vægt 257.1426