missing translation for 'onlineSavingsMsg'
Learn More

Desdiacetyl-8-oxo Famciclovir-d4, TRC

Product Code. 30367686
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30367686 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30367686 Supplier Toronto Research Chemicals Supplier No. TRCD2889321MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Desdiacetyl-8-oxo Famciclovir-d4
CAS 1346603-55-1
Molekylær formel C10H11D4N5O3
Mængde 1 mg
Synonym 2-Amino-7,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-8H-purin-8-one-d4, BRL 48959-d4, 8-Oxo-6-deoxypenciclovir-d4
InChI formel InChI=1S/C10H15N5O3/c11-9-12-3-7-8(14-9)15(10(18)13-7)2-1-6(4-16)5-17/h3,6,16-17H,1-2,4-5H2,(H,13,18)(H2,11,12,14)/i1D2,2D2
SMIL [2H]C([2H])(C(CO)CO)C([2H])([2H])N1C(=NC2=CNC(=N)N=C12)O
IUPAC navn 2-[1,1,2,2-tetradeuterio-2-(8-hydroxy-2-imino-1H-purin-9-yl)ethyl]propane-1,3-diol
Molekylvægt (g/mol) 257.28
Formel vægt 257.1426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.