Få mere at vide
Deschlorodesloratadine Hydrochloride (6,11-Dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine Hydrochloride), Mikromol™
Artikelnummer. 30462145
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462145 4 x 25 mg
17963516 25 mg
17964026 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30462145 Leverandør Mikromol Leverandørnr. MM0257.1900254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Deschlorodesloratadine Hydrochloride (6,11-Dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deschlorodesloratadine Hydrochloride
CAS 2172059-46-8
Analyt- eller komponentnavne Deschlorodesloratadine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H20 N2 . Cl H
Certificeringer/compliance ISO 9001
Synonym Deschlorodesloratadine Hydrochloride, 6,11-Dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine Hydrochloride
InChI formel InChI=1S/C19H20N2.ClH/c1-2-6-17-14(4-1)7-8-16-5-3-11-21-19(16)18(17)15-9-12-20-13-10-15;/h1-6,11,20H,7-10,12-13H2;1H
Løsningstype Neat
SMIL Cl.C1CC(=C2c3ccccc3CCc4cccnc24)CCN1
IUPAC navn 11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine;hydrochloride
Molekylvægt (g/mol) 312.84
Mængde 4 x 25 mg
Formel vægt 312.1393 g/mol