Få mere at vide
Deschloro Clomiphene Hydrochloride (E/Z Mixture), TRC
Artikelnummer. 30348974
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30348974 1 mg
30368050 5 mg
30364973 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30348974 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2889811MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 74056-26-1
Molekylær formel C26 H29 N O . Cl H
Molekylvægt (g/mol) 407.98
Synonym 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine hydrochloride (1:1), Ethanamine, 2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, hydrochloride (9CI), Triethylamine, 2-[p-(1,2-diphenylvinyl)phenoxy]-, hydrochloride, (E,Z)-2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride, 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine Hydrochloride, Clomiphene Related Compound A (USP)
IUPAC navn 2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine;hydrochloride
SMIL Cl.CCN(CC)CCOc1ccc(cc1)C(=Cc2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale (E,Z)-2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C26H29NO.ClH/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22;/h5-18,21H,3-4,19-20H2,1-2H3;1H
Formel vægt 407.2016