Få mere at vide
Deschloro Clomiphene-d5, TRC
Artikelnummer. 30365075
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30365075 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365075 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2889825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deschloro Clomiphene-d5
CAS 1346606-17-4
Molekylær formel C26H24D5NO
Mængde 5 mg
Synonym 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine-d5
InChI formel InChI=1S/C26H29NO/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22/h5-18,21H,3-4,19-20H2,1-2H3/b26-21-/i5D,7D,8D,11D,12D
SMIL [2H]c1c([2H])c([2H])c(\C=C(\c2ccccc2)/c3ccc(OCCN(CC)CC)cc3)c([2H])c1[2H]
IUPAC navn N,N-diethyl-2-[4-[(Z)-2-(2,3,4,5,6-pentadeuteriophenyl)-1-phenylethenyl]phenoxy]ethanamine
Molekylvægt (g/mol) 376.55
Formel vægt 376.2563