Få mere at vide
Desbutyl Lumefantrine-d9 (E/Z-Mixture), TRC
Artikelnummer. 30359680
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30359680 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30359680 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28889710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desbutyl Lumefantrine-d9 (E/Z-Mixture)
CAS 1346606-35-6
Molekylær formel C26H15D9Cl3NO
Mængde 10 mg
Synonym α-[(Butylamino-d9)methyl]-2,7-dichloro-9-[(4-chlorophenyl)methylene]-9H-fluorene-4-methanol
InChI formel InChI=1S/C26H24Cl3NO/c1-2-3-10-30-15-25(31)24-14-19(29)13-23-21(11-16-4-6-17(27)7-5-16)22-12-18(28)8-9-20(22)26(23)24/h4-9,11-14,25,30-31H,2-3,10,15H2,1H3/b21-11-/i1D3,2D2,3D2,10D2
SMIL ClC(C=C1)=CC=C1/C=C2C3=CC(Cl)=CC(C(CNC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])O)=C3C4=C\2C=C(Cl)C=C4
Molekylvægt (g/mol) 481.89
Formel vægt 480.1488