Få mere at vide
Desamino Glufosinate-d3, TRC
Artikelnummer. 30353804
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30353804 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353804 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2884621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desamino Glufosinate-d3
CAS 1794938-71-8
Molekylær formel C5H8D3O4P
Mængde 1 mg
Synonym 4-(Hydroxy-(trideuteriomethyl)phosphoryl)butanoic acid
InChI formel InChI=1S/C5H11O4P/c1-10(8,9)4-2-3-5(6)7/h2-4H2,1H3,(H,6,7)(H,8,9)/i1D3
SMIL C(CCC(O)=O)P(C([2H])([2H])[2H])(=O)O
IUPAC navn 4-(hydroxy(methyl-d3)phosphoryl)butanoic acid
Molekylvægt (g/mol) 169.13
Formel vægt 169.06