missing translation for 'onlineSavingsMsg'
Learn More

Desamino Chloroimiquimod, TRC

Product Code. 30361535
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30361535 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361535 Supplier Toronto Research Chemicals Supplier No. TRCD2882955MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 4-Chloro-1-isobutyl-1H-imidazo[4,5-c]quinoline
CAS 99010-64-7
Molekylær formel C14 H14 Cl N3
Mængde 5 mg
Synonym 1H-Imidazo[4,5-c]quinoline, 4-chloro-1-(2-methylpropyl)-, 4-Chloro-1-isobutyl-1H-imidazo[4,5-c]quinoline, NSC 672965
InChI formel InChI=1S/C14H14ClN3/c1-9(2)7-18-8-16-12-13(18)10-5-3-4-6-11(10)17-14(12)15/h3-6,8-9H,7H2,1-2H3
SMIL CC(C)Cn1cnc2c(Cl)nc3ccccc3c12
IUPAC navn 4-chloro-1-(2-methylpropyl)imidazo[4,5-c]quinoline
Molekylvægt (g/mol) 259.73
Formel vægt 259.0876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.