Få mere at vide
Desallyl Rocuronium Diacetate, TRC
Artikelnummer. 30374670
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30374670 10 mg
1 options
Artikelnummer. 30374670 Leverandør Toronto Research Chemicals Leverandørnr. TRCD99101010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desallyl Rocuronium Diacetate
CAS 119302-22-6
Molekylær formel C31H50N2O5
Mængde 10 mg
Synonym Androstane-3,17-diol, 2-(4-morpholinyl)-16-(1-pyrrolidinyl)-, 3,17-diacetate, (2β,3α,5α,16β,17β)-, Androstane-3,17-diol, 2-(4-morpholinyl)-16-(1-pyrrolidinyl)-, diacetate (ester), (2β,3α,5α,16β,17β)- (9CI), (2β,3α,5α,16β,17β)-2-(4-Morpholinyl)-16-(1-pyrrolidinyl)androstane-3,17-diol diacetate, (2β,3α,5α,16β,17β)-3,17-Diacetoxy-2-(4-morpholinyl)-16-(1-pyrrolidinyl)androstane, 2β-(4-Morpholinyl)-16β-(1-pyrrolidinyl)-5α-androstane-3α,17β-diol diacetate, 2β-(Morpholin-4-yl)-16β-(pyrrolidin-1-yl)-5α-androstane-3α,17β-diol 3,17-Diacetate
InChI formel InChI=1S/C31H50N2O5/c1-20(34)37-28-17-22-7-8-23-24(31(22,4)19-27(28)33-13-15-36-16-14-33)9-10-30(3)25(23)18-26(29(30)38-21(2)35)32-11-5-6-12-32/h22-29H,5-19H2,1-4H3/t22-,23+,24-,25-,26-,27-,28-,29-,30-,31-/m0/s1
SMIL C[C@@]12[C@]([C@]3([C@](CC1)([C@]4(C)[C@@](CC3)(C[C@H](OC(C)=O)[C@H](C4)N5CCOCC5)[H])[H])[H])(C[C@@H]([C@@H]2OC(C)=O)N6CCCC6)[H]
Molekylvægt (g/mol) 530.74