Få mere at vide
Desalkyl Ebastine-d5, TRC
Artikelnummer. 30351309
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30351309 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30351309 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28877210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desalkyl Ebastine-d5
CAS 1189424-36-9
Molekylær formel C18H16D5NO
Mængde 10 mg
Synonym 4-(Diphenylmethoxy-d5)piperidine, 4-(Benzhydryloxy-d5)piperidine
InChI formel InChI=1S/C18H21NO/c1-3-7-15(8-4-1)18(16-9-5-2-6-10-16)20-17-11-13-19-14-12-17/h1-10,17-19H,11-14H2/i1D,3D,4D,7D,8D
SMIL [2H]C(C([2H])=C([2H])C([2H])=C1[2H])=C1C(OC2CCNCC2)C3=CC=CC=C3
Molekylvægt (g/mol) 272.4