Få mere at vide
Desacetyllanatoside C, TRC
Artikelnummer. 30134757
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134757 5 mg
30138016 100 mg
30012599 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134757 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2883205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17598-65-1
Molekylær formel C47 H74 O19
Molekylvægt (g/mol) 943.08
Synonym 3β-[(O-β-d-glucopyranosyl-(1→4)-O-2,6-dideoxy-β-d-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-d-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-d-ribo-hexopyranosyl)oxy]-12β,14-dihydroxy-5β,14β-card-20(22)-enolide, Card-20(22)-enolide, 3-[(O-β-D-glucopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxy-, (3β,5β,12β)-, Lanatoside C, deacetyl- (6CI,7CI,8CI), (3β,5β,12β)-3-[(O-β-D-Glucopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide, Cedilanid D, De-O-acetyllanatoside C, Deacetylanatoside C, Deacetyllanatoside C, Desace, Desacetyllanatoside C, Desaci, Deslanatoside C, Deslanoside, Deslanoside C, Purpurea glycoside C
IUPAC navn 3-[(3S,5R,8R,9S,10S,12R,13S,14S,17R)-12,14-dihydroxy-3-[(2R,4S,5S,6R)-4-hydroxy-5-[(2S,4S,5S,6R)-4-hydroxy-5-[(2S,4S,5S,6R)-4-hydroxy-6-methyl-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-methyloxan-2-yl]oxy-6-methyloxan-2-yl]oxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
SMIL C[C@H]1O[C@H](C[C@H](O)[C@@H]1O[C@H]2C[C@H](O)[C@H](O[C@H]3C[C@H](O)[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@@H](C)O3)[C@@H](C)O2)O[C@H]5CC[C@@]6(C)[C@H](CC[C@@H]7[C@@H]6C[C@@H](O)[C@]8(C)[C@H](CC[C@]78O)C9=CC(=O)OC9)C5
Tekniske data
Kemisk navn eller materiale Deacetyllanatoside C
Mængde 5 mg
InChI formel InChI=1S/C47H74O19/c1-20-41(64-36-16-30(50)42(21(2)60-36)65-37-17-31(51)43(22(3)61-37)66-44-40(56)39(55)38(54)32(18-48)63-44)29(49)15-35(59-20)62-25-8-10-45(4)24(13-25)6-7-27-28(45)14-33(52)46(5)26(9-11-47(27,46)57)23-12-34(53)58-19-23/h12,20-22,24-33,35-44,48-52,54-57H,6-11,13-19H2,1-5H3/t20-,21-,22-,24-,25+,26-,27-,28+,29+,30+,31+,32-,33-,35+,36+,37+,38-,39+,40-,41-,42-,43-,44+,45+,46+,47+/m1/s1
Formel vægt 942.4824