Få mere at vide
Desacetyl Diltiazem-d3, TRC
Artikelnummer. 30365315
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30365315 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365315 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28868710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desacetyl Diltiazem-d3
CAS 1246815-32-6
Molekylær formel C20H21D3N2O3S
Mængde 10 mg
Synonym (2S-cis)-5-[(Dimethylamino-d3)ethyl]-2,3-dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one, Deacetyl-d-diltiazem-d3, Deacetyldiltiazem-d3
InChI formel InChI=1S/C20H24N2O3S/c1-21(2)12-13-22-16-6-4-5-7-17(16)26-19(18(23)20(22)24)14-8-10-15(25-3)11-9-14/h4-11,18-19,23H,12-13H2,1-3H3/t18-,19+/m1/s1/i1D3
SMIL O[C@@H](C(N(CCN(C)C([2H])([2H])[2H])C(C=CC=C1)=C1S2)=O)[C@@H]2C3=CC=C(OC)C=C3
Molekylvægt (g/mol) 375.5