Få mere at vide
Des(sulfonylethyl)amino Hydroxy Lapatinib, TRC
Artikelnummer. 30365831
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365831 10 mg
30356070 2.5 mg
30358998 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365831 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29742310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 320337-48-2
Molekylær formel C26H19ClFN3O3
Molekylvægt (g/mol) 475.9
Synonym 5-[4-[[3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]amino]-6-quinazolinyl]-2-furanmethanol
SMIL OCc1oc(cc1)c2ccc3ncnc(Nc4ccc(OCc5cccc(F)c5)c(Cl)c4)c3c2
Tekniske data
Kemisk navn eller materiale Des(sulfonylethyl)amino Hydroxy Lapatinib
Mængde 10 mg
InChI formel InChI=1S/C26H19ClFN3O3/c27-22-12-19(5-8-25(22)33-14-16-2-1-3-18(28)10-16)31-26-21-11-17(4-7-23(21)29-15-30-26)24-9-6-20(13-32)34-24/h1-12,15,32H,13-14H2,(H,29,30,31)