Få mere at vide
Des-N-(methoxycarbonyl)-L-tert-leucine Atazanavir Trihydrochloride, TRC
Artikelnummer. 30352991
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30352991 50 mg
30360432 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30352991 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29032050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Des-N-(methoxycarbonyl)-L-tert-leucine Atazanavir Trihydroch
CAS 198904-87-9
Molekylær formel C22 H26 N4 O . 3 Cl H
Mængde 50 mg
Synonym Des-N-(methoxycarbonyl)-L-tert-leucine Atazanavir Trihydrochloride, (αS,βS)-β-amino-α-[[1-[[4-(2-pyridinyl)phenyl]methyl]hydrazinyl]methyl]-benzenepropanol trihydrochloride, (αS,βS)-β-amino-α-[[1-[[4-(2-pyridinyl)phenyl]methyl]hydrazinyl]methyl]-benzenepropanol hydrochloride (1:3), [S-(R*,R*)]-β-Amino-α-[[1-[[4-(2-pyridinyl)phenyl]methyl]hydrazino]methyl]-benzenepropanol, (2S,3S)-3-Amino-1-[amino-[[4-(2-pyridyl)phenyl]methyl]amino]-4-phenylbutan-2-ol Trihydrochloride
InChI formel InChI=1S/C22H26N4O.3ClH/c23-20(14-17-6-2-1-3-7-17)22(27)16-26(24)15-18-9-11-19(12-10-18)21-8-4-5-13-25-21;;;/h1-13,20,22,27H,14-16,23-24H2;3*1H/t20-,22-;;;/m0.../s1
SMIL Cl.Cl.Cl.N[C@@H](Cc1ccccc1)[C@@H](O)CN(N)Cc2ccc(cc2)c3ccccn3
IUPAC navn (2S,3S)-3-amino-1-[amino-[(4-pyridin-2-ylphenyl)methyl]amino]-4-phenylbutan-2-ol;trihydrochloride
Molekylvægt (g/mol) 471.85
Formel vægt 470.1407