Få mere at vide
Des-methylformate Eplerenone, TRC
Artikelnummer. 30365276
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30365276 2.5 mg 2.5mg
30352292 25 mg -
30356282 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365276 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2939552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50394-27-9
Molekylær formel C22H28O4
Molekylvægt (g/mol) 356.46
Synonym 9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-gamma-lactone-pr50394-27-9egn-4-ene-21-carboxylic Acid, SC 12590
SMIL C[C@]12C[C@H]3O[C@@]34[C@@H](CCC5=CC(=O)CC[C@]45C)[C@@H]1CC[C@@]26CCC(=O)O6
Tekniske data
Kemisk navn eller materiale Des-methylformate Eplerenone
Mængde 2.5 mg
InChI formel InChI=1S/C22H28O4/c1-19-8-5-14(23)11-13(19)3-4-16-15-6-9-21(10-7-18(24)26-21)20(15,2)12-17-22(16,19)25-17/h11,15-17H,3-10,12H2,1-2H3
Formel vægt 356.1988