Få mere at vide
Des-4'(methylpiperidine-4-carboxamide)-4'-hydroxymethyl Revefenacin, TRC
Artikelnummer. 30364458
Skift visning
Mængde:
2.5 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364458 2.5 mg
30360873 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364458 Leverandør Toronto Research Chemicals Leverandørnr. TRCD1198602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Des-4'(methylpiperidine-4-carboxamide)-4'-hydroxymethyl Revefenacin
CAS 1682660-39-4
Molekylær formel C29 H33 N3 O4
Mængde 2.5 mg
Synonym 1-(2-(4-(Hydroxymethyl)-N-methylbenzamido)ethyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamate
InChI formel InChI=1S/C29H33N3O4/c1-31(28(34)24-13-11-22(21-33)12-14-24)19-20-32-17-15-25(16-18-32)36-29(35)30-27-10-6-5-9-26(27)23-7-3-2-4-8-23/h2-14,25,33H,15-21H2,1H3,(H,30,35)
SMIL CN(CCN1CCC(CC1)OC(=O)Nc2ccccc2c3ccccc3)C(=O)c4ccc(CO)cc4
IUPAC navn [1-[2-[[4-(hydroxymethyl)benzoyl]-methylamino]ethyl]piperidin-4-yl] N-(2-phenylphenyl)carbamate
Molekylvægt (g/mol) 487.59
Formel vægt 487.247