Få mere at vide
Des-2-methyl-2-hydroxymethyl Metalaxy, TRC
Artikelnummer. 30476229
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476229 250 mg
30468609 25 mg
30478289 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476229 Leverandør Toronto Research Chemicals Leverandørnr. TRCM881235250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85933-49-9
Molekylær formel C15 H21 N O5
Molekylvægt (g/mol) 295.33
Synonym Alanine, N-[2-(hydroxymethyl)-6-methylphenyl]-N-(methoxyacetyl)-, methyl ester, DL-Alanine, N-[2-(hydroxymethyl)-6-methylphenyl]-N-(methoxyacetyl)-, methyl ester
IUPAC navn methyl 2-[2-(hydroxymethyl)-N-(2-methoxyacetyl)-6-methylanilino]propanoate
SMIL COCC(=O)N(C(C)C(=O)OC)c1c(C)cccc1CO
Tekniske data
Kemisk navn eller materiale Metalaxyl-hydroxymethyl
Mængde 250 mg
InChI formel InChI=1S/C15H21NO5/c1-10-6-5-7-12(8-17)14(10)16(13(18)9-20-3)11(2)15(19)21-4/h5-7,11,17H,8-9H2,1-4H3
Formel vægt 295.142