Learn More
Dermofungin, TRC
Product Code. 30136156
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136156 10 g 10g
30133713 50 g 50g
30007228 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136156 Supplier Toronto Research Chemicals Supplier No. TRCD28193010G
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 130-16-5
Molecular Formula C9 H6 Cl N O
Molecular Weight (g/mol) 179.6
Synonym Cloxiquine, 5-Chloro-8-hydroxyquinoline, 5-Chloro-8-oxyquinoline, 5-Chloro-8-quinolinol, 5-Chloroquinoline-8-ol, 5-Chlorooxine, 8-Hydroxy-5-chloroquinoline, Cloxyquin, Dermofongin, Dermofungin, NSC 35083, NSC 53182, NSC 74943, Clioquinol Imp. A (EP)
IUPAC Name 5-chloroquinolin-8-ol
SMILES Oc1ccc(Cl)c2cccnc12
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H6ClNO/c10-7-3-4-8(12)9-6(7)2-1-5-11-9/h1-5,12H
Formula Weight 179.0138
Percent Purity >95%
Chemical Name or Material Cloxiquine
Grade HPLC