Få mere at vide
Dequalinium Chloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dequalinium chloride |
| CAS | 522-51-0 |
| Analyt- eller komponentnavne | Dequalinium chloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C30 H40 N4 . 2 Cl |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Dequalinium chloride, 1,1'-(decane-1,10-diyl)bis(4-amino-2-methylquinolinium) dichloride, 1,1'-Decamethylenebis[4-aminoquinaldinium chloride], Quinaldinium, 1,1'-decamethylenebis[4-amino-, dichloride, Quinolinium, 1,1'-(1,10-decanediyl)bis[4-amino-2-methyl-, dichloride, BADQ 10, BAQD 10, Decamethylenebis[4-aminoquinaldinium chloride], Decamine, Decamine (pharmaceutical), Decatylen, Dekadin, Dekamiln, Dekamin, Dequacets, Dequadin, Dequadin chloride, Dequafungen, Dequalinium chloride, Dequavagyn, Dequavet, Efisol, Eriosept, Evazol, Fluomycin N, Grocreme, Ivazil, LDN 0096422, Labosept, NSC 166454, Optipect, Oralgol, Phylletten, Polycidine, Rumilet, Sanoral, Sentril, Sorot |
| InChI formel | InChI=1S/C30H38N4.2ClH/c1-23-21-27(31)25-15-9-11-17-29(25)33(23)19-13-7-5-3-4-6-8-14-20-34-24(2)22-28(32)26-16-10-12-18-30(26)34;;/h9-12,15-18,21-22,31-32H,3-8,13-14,19-20H2,1-2H3;2*1H |
| Løsningstype | Neat |
| SMIL | [Cl-].[Cl-].Cc1cc(N)c2ccccc2[n+]1CCCCCCCCCC[n+]3c(C)cc(N)c4ccccc34 |
| Vis mere |