Få mere at vide
Depropylamino Hydroxy Propafenone-d5, TRC
Artikelnummer. 30352203
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30352203 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352203 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2886372.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Depropylamino Hydroxy Propafenone-d5
CAS 1346598-59-1
Molekylær formel C18H15D5O4
Mængde 2.5 mg
Synonym 1-[2-[2,3-Dihydroxy(propoxy-d5)]phenyl]-3-phenyl-1-propanone
InChI formel InChI=1S/C18H20O4/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14/h1-9,15,19-20H,10-13H2/i12D2,13D2,15D
SMIL [2H]C([2H])(O)C([2H])(O)C([2H])([2H])Oc1ccccc1C(=O)CCc2ccccc2
IUPAC navn 1-[2-(1,1,2,3,3-pentadeuterio-2,3-dihydroxypropoxy)phenyl]-3-phenylpropan-1-one
Molekylvægt (g/mol) 305.38
Formel vægt 305.1675