missing translation for 'onlineSavingsMsg'
Få mere at vide

Depropylamino Hydroxy Propafenone-d5, TRC

Artikelnummer. 30353000
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30353000 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353000 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28863725MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Depropylamino Hydroxy Propafenone-d5
CAS 1346598-59-1
Molekylær formel C18H15D5O4
Mængde 25 mg
Synonym 1-[2-[2,3-Dihydroxy(propoxy-d5)]phenyl]-3-phenyl-1-propanone
InChI formel InChI=1S/C18H20O4/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14/h1-9,15,19-20H,10-13H2/i12D2,13D2,15D
SMIL [2H]C([2H])(O)C([2H])(O)C([2H])([2H])Oc1ccccc1C(=O)CCc2ccccc2
IUPAC navn 1-[2-(1,1,2,3,3-pentadeuterio-2,3-dihydroxypropoxy)phenyl]-3-phenylpropan-1-one
Molekylvægt (g/mol) 305.38
Formel vægt 305.1675
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.