Få mere at vide
Depropylamino Chloro Propafenone, TRC
Artikelnummer. 30362208
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362208 50 mg
30360212 100 mg
30352838 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362208 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28865550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 165279-79-8
Molekylær formel C18 H19 Cl O3
Molekylvægt (g/mol) 318.79
Synonym 1-[2-[(2RS)-3-Chloro-2-hydroxypropoxy]phenyl]-3-phenylpropan-1-one, Propafenone Hydrochloride Imp. E (EP)
IUPAC navn 1-[2-(3-chloro-2-hydroxypropoxy)phenyl]-3-phenylpropan-1-one
SMIL OC(CCl)COc1ccccc1C(=O)CCc2ccccc2
Tekniske data
Kemisk navn eller materiale 1-[2-[(2RS)-3-Chloro-2-hydroxypropoxy]phenyl]-3-phenylpropan-1-one
Mængde 50 mg
InChI formel InChI=1S/C18H19ClO3/c19-12-15(20)13-22-18-9-5-4-8-16(18)17(21)11-10-14-6-2-1-3-7-14/h1-9,15,20H,10-13H2
Formel vægt 318.1023