missing translation for 'onlineSavingsMsg'
Få mere at vide

Deoxyviolacein, TRC

Artikelnummer. 30377111
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30377111 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377111 Supplier Toronto Research Chemicals Supplier No. TRCD4560132.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Deoxyviolacein
CAS 5839-61-2
Molekylær formel C20 H13 N3 O2
Mængde 2.5 mg
Synonym (3E)-3-[1,2-Dihydro-5-(1H-indol-3-yl)-2-oxo-3H-pyrrol-3-ylidene]-1,3-dihydro-2H-indol-2-one2H-Indol-2-one
InChI formel InChI=1S/C20H13N3O2/c24-19-13(18-12-6-2-4-8-16(12)22-20(18)25)9-17(23-19)14-10-21-15-7-3-1-5-11(14)15/h1-10,21H,(H,22,25)(H,23,24)/b18-13+
SMIL O=C1NC(=C/C/1=C/2\C(=O)Nc3ccccc23)c4c[nH]c5ccccc45
IUPAC navn (3E)-3-[5-(1H-indol-3-yl)-2-oxo-1H-pyrrol-3-ylidene]-1H-indol-2-one
Molekylvægt (g/mol) 327.336
Formel vægt 327.101
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.