Få mere at vide
Deoxynojirimycin, TRC
Artikelnummer. 30133680
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133680 10 mg
30133414 100 mg
17979074 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133680 Leverandør Toronto Research Chemicals Leverandørnr. TRCD24500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 19130-96-2
Molekylær formel C6 H13 N O4
Molekylvægt (g/mol) 163.17
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, (2R,3R,4R,5S)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, [2R-(2α,3β,4α,5β)]-, Glucopyranose, 5-amino-1,5-dideoxy-, D- (8CI), (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol, 1,5-Dideoxy-1,5-imino-D-glucitol, 1-Deoxynojirimycin, 5-Amino-1,5-dideoxy-D-glucopyranose, BAY-h 5595, D-1-Deoxynojirimycin, D-Glucitol, 1,5-dideoxy-1,5-imino-, Deoxynojirimycin, Duvoglustat, Moranolin, Moranoline, Nojirimycin, 1-deoxy-
IUPAC navn (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
SMIL OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Deoxynojirimycin
Mængde 10 mg
InChI formel InChI=1S/C6H13NO4/c8-2-3-5(10)6(11)4(9)1-7-3/h3-11H,1-2H2/t3-,4+,5-,6-/m1/s1
Formel vægt 163.0845