Få mere at vide
Deoxygalactonojirimycin-15N Hydrochloride, TRC
Artikelnummer. 30364330
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364330 1 mg
30361146 10 mg
30362005 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30364330 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2365021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219134-37-8
Molekylær formel C6H14Cl15NO4
Molekylvægt (g/mol) 200.63
Synonym (2R,3S,4R,5S)-2-Hydroxymethyl-3,4,5-piperidinetriol-15N Hydrochloride, Migalastat-15N Hydrochloride
SMIL Cl.OC[C@H]1[15NH]C[C@H](O)[C@@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Deoxygalactonojirimycin-15N Hydrochloride
Mængde 1 mg
InChI formel InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H
Formel vægt 200.0582