missing translation for 'onlineSavingsMsg'
Få mere at vide

Deoxydihydroartemisinin (Alpha,Beta Mixture), TRC

Artikelnummer. 30364380
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364380 100 mg
30349152 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30364380 Leverandør Toronto Research Chemicals Leverandørnr. TRCD239660100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 72807-92-2
Molekylær formel C15H24O4
Molekylvægt (g/mol) 268.35
Synonym 2-Deoxydihydroartemisinin, Chemal 330, Deoxydihydroartemisinin, Desoxydihydroartemisinin, Dihydrodeoxyartemisinin, [3R-(3R,3aS,6R,6aS,9S,10aR,10bR)]-[Partial]-decahydro-3,6,9-trimethyl-10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2-ol, (3R,3aS,6R,6aS,9S,10aR,10bR)-Decahydro-3,6,9-trimethyl-10aH-9,10b-Epoxypyrano[4,3,2-jk][2]benzoxepin-2-ol
SMIL C[C@@]1([H])C(O)O[C@]2([H])[C@@]34[C@](CC[C@@](C)(O4)O2)([H])[C@H](C)CC[C@]31[H]

Tekniske data

Kemisk navn eller materiale Deoxydihydroartemisinin (alpha,beta Mixture)
Mængde 100 mg
InChI formel InChI=1S/C15H24O4/c1-8-4-5-11-9(2)12(16)17-13-15(11)10(8)6-7-14(3,18-13)19-15/h8-13,16H,4-7H2,1-3H3/t8-,9-,10+,11+,12?,13+,14-,15-/m1/s1
Formel vægt 268.1675
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.