Beskrivelse
Deoxycytidylate deaminase Polyclonal specifically detects Deoxycytidylate deaminase in Human samples. It is validated for Western Blot.
Tekniske data
Tekniske data
| Antigen | Deoxycytidylate deaminase |
| Applikationer | Western Blot |
| Klassifikation | Polyclonal |
| Koncentration | 0.5 mg/ml |
| Konjugeret | Unconjugated |
| Fortynding | Western Blot 1.0 ug/ml |
| Formulering | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Genadgangsnr. | D3DP49 |
| Gene Alias | dCMP deaminaseEC 3.5.4.12, deoxycytidylate deaminase, MGC111062 |
| Gensymboler | DCTD |
| Vis mere |