Få mere at vide
Deoxycholic Acid 3-O-Sulfate Disodium Salt, TRC
Artikelnummer. 30366725
Skift visning
Mængde:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366725 2.5 mg
30361711 10 mg
30361909 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366725 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2327152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60237-35-6
Molekylær formel C24H38Na2O7S
Molekylvægt (g/mol) 516.6
Synonym 12-Hydroxy-3-(sulfooxy)-(3alpha,5beta,12alpha)-cholan-24-oic Acid
SMIL [Na+].[Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C)OS(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale Deoxycholic Acid 3-O-Sulfate Disodium Salt
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C24H40O7S.2Na/c1-14(4-9-22(26)27)18-7-8-19-17-6-5-15-12-16(31-32(28,29)30)10-11-23(15,2)20(17)13-21(25)24(18,19)3;;/h14-21,25H,4-13H2,1-3H3,(H,26,27)(H,28,29,30);;/q;2*+1/p-2/t14-,15-,16-,17?,18-,19+,20?,21+,23+,24-;;/m1../s1
Formel vægt 516.2134
Procent renhed >95%