Få mere at vide
Deoxycholic-2,2,4,4-d4 Acid, TRC
Artikelnummer. 30349634
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30349634 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349634 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2326485MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deoxycholic Acid (2,2,4,4-D4)
CAS 112076-61-6
Molekylær formel C24H36D4O4
Mængde 5 mg
Synonym Cholan-24-oic-2,2,4,4-d4 acid, 3,12-dihydroxy-, (3α,5β,12α)-, (3α,5β,12α)-3,12-Dihydroxycholan-24-oic-2,2,4,4-d4 acid, Deoxycholic acid-d4, Deoxycholic-2,2,4,4-d4 acid, [2,2,4,4-2H4]-Deoxycholic acid, Deoxycholic acid (2,2,4,4-D4)
InChI formel InChI=1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1/i10D2,12D2
SMIL C[C@@]12[C@]([C@]3([C@@]([C@]4(C)[C@](CC3)(C([C@H](O)C(C4)([2H])[2H])([2H])[2H])[H])(C[C@@H]1O)[H])[H])(CC[C@@]2([C@@H](CCC(O)=O)C)[H])[H]
IUPAC navn (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl-2,2,4,4-d4)pentanoic acid
Molekylvægt (g/mol) 396.6
Formel vægt 396.3178